missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BCAP29 Partial ORF (AAH08478, 125 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055973-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: TQLAKELSNKGVLKTQAENTNKAAKKFMEENEKLKRILKSHGKDEECVLEAENKKLVEDQEKLKTELRKTSDALSKAQNDVMEMKMQSERLSKEYDQLLKEHSELQDRLERGNKKRLSpecifications
AAH08478 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.50kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TQLAKELSNKGVLKTQAENTNKAAKKFMEENEKLKRILKSHGKDEECVLEAENKKLVEDQEKLKTELRKTSDALSKAQNDVMEMKMQSERLSKEYDQLLKEHSELQDRLERGNKKRL | |
RUO | |
BCAP29 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55973 | |
BCAP29 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAP29/DKFZp686M2086 | |
BCAP29 | |
Recombinant | |
wheat germ expression system |