missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCAP29 Full-length ORF (NP_061332.2, 1 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055973-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192003

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

Sequence: MTLQWAAVATFLYAEIGLILIFCLPFIPPQRWQKIFSFNVWGKIATFWNKAFLTIIILLIVLFLDAVREVRKYSSVHTIEKSSTSRPDAYEHTQMKLFRSQRNLYISGFSLFFWLVLRRLVTLITQLAKELSNKGVLKTQAENTNKAAKKFMEENEKLKRILKSHGKDEECVLEAENKKLVEDQEKLKTELRKTSDALSKAQNDVMEMKMQSERLSKEYDQLLKEHSELQDRLERGNKKRL
Specifications

Specifications

NP_061332.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
BCAP29
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
55973
BCAP29 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAP29/DKFZp686M2086
BCAP29
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers