Learn More
Abnova™ Human BAI3 Partial ORF (NP_001695, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000577-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis. [provided by RefSeq]
Sequence: QDFWCSTLVKGVIYGSYSVSEMFPKNFTNCTWTLENPDPTKYSIYLKFSKKDLSCSNFSLLAYQFDHFSHEKIKDLLRKNHSIMQLCNSKNAFVFLQYDKNFIQIRRVFPSpecifications
NP_001695 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QDFWCSTLVKGVIYGSYSVSEMFPKNFTNCTWTLENPDPTKYSIYLKFSKKDLSCSNFSLLAYQFDHFSHEKIKDLLRKNHSIMQLCNSKNAFVFLQYDKNFIQIRRVFP | |
RUO | |
BAI3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
577 | |
BAI3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0550/MGC133100 | |
BAI3 | |
Recombinant | |
wheat germ expression system |