Learn More
ACROBiosystems Human B7-H5 / Gi24 / VISTA Protein, Fc Tag
Brand: ACROBiosystems B75-H5259-1MG
Additional Details : Weight : 0.00001kg
Description
Platelet receptor Gi24, also known as B7-H5 and stress-induced secreted protein-1 (Sisp-1), is a protein that in humans is encoded by the C10orf54 gene, which contains 1 Ig-like (immunoglobulin-like) domain. As for C10orf54 gene, C10orf54 appears to positively interact with BMP-4, potentiating BMP signaling and the transition from an undifferentiated to a differentiated state on ESCs. Human C10orf54 undergoes proteolytic cleavage by MT1-MMP, generating a soluble 30 kDa extracellular fragment plus a 25-30 kDa membrane-bound fragment.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifications
AAH20568 | |
1 mg | |
Phe 33 - Ala 194 | |
B7-H5,SISP1,Gi24,VISTA | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
44.8 kDa | |
Human B7-H5, Fc Tag (B75-H5259) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568). | |
-20°C | |
B7-H5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |