Learn More
Proteintech™ Human B7-H3/CD276 ELISA Kit
Product Code. 16801556 Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Product Code. 16801556
Brand: Proteintech™ KE0014396T
Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous B7-H3/CD276
  • Recognizes human B7-H3/CD276
  • B7-H3 (CD276) is a Type I transmembrane protein expressed on many tissues and cell types
    • B7-H3 is a 100-kDa glycoprotein belonging to B7 immunoregulatory family
      • Participates in regulation of T-cell-mediated immune response by functioning as T cell costimulator and coinhibitor
    • Overexpressed on human solid cancers, implicated in cancer progression and metastasis, target for cancer immunotherapy
Process:
  • Antibody specific for B7-H3/CD276 pre-coated onto microwells
  • B7-H3/CD276 protein in samples captured by the coated antibody after incubation
  • After extensive washing, another horseradish peroxidase (HRP)-conjugated antibody specific for B7-H3/CD276 is added to detect captured B7-H3/CD276 protein
  • For signal development, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Accession Number BC062581
Assay Range 9.4 to 600 pg/mL
Assay Sensitivity 0.6 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 80381
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID Q5ZPR3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human TNFRSF9 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine