Learn More
Abnova™ Human B4GALT7 Partial ORF (NP_009186, 194 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011285-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme encoded by this gene attaches the first galactose in the common carbohydrate-protein (GlcA-beta1,3-Gal-beta1,3-Gal-beta1,4-Xyl-beta1-O-Ser) linkage found in proteoglycans. Manganese is required as a cofactor. This enzyme differs from the other six beta4GalTs because it lacks the conserved beta4GalT1-beta4GalT6 Cys residues and it is located in cis-Golgi instead of trans-Golgi. Two single-nucleotide mutations were identified from a patient with the progeroid type of Ehlers-Danlos syndrome. [provided by RefSeq]
Sequence: YHYKTYVGGILLLSKQHYRLCNGMSNRFWGWGREDDEFYRRIKGAGLQLFRPSGITTGYKTFRHLHDPAWRKRDQKRIAAQKQEQFKVDREGGLNTVKYHSpecifications
NP_009186 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YHYKTYVGGILLLSKQHYRLCNGMSNRFWGWGREDDEFYRRIKGAGLQLFRPSGITTGYKTFRHLHDPAWRKRDQKRIAAQKQEQFKVDREGGLNTVKYH | |
RUO | |
B4GALT7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11285 | |
B4GALT7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B4GAL-T7/XGALT-1/XGALT1/XGPT1/beta4Gal-T7 | |
B4GALT7 | |
Recombinant | |
wheat germ expression system |