missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AUTS2 Partial ORF (NP_056385, 108 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026053-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125286

  • 359.00€ / 10µg

Estimated Shipment: 13-09-2022
to see stock.
Add to basket

Description

Description

Sequence: LKPQERVEKRQTPLTKKKREALTNGLSFHSKKSRLSHPHHYSSDRENDRNLCQHLGKRKKMPKALRQLKPGQNSCRDSDSESASGESKGFH
Specifications

Specifications

NP_056385
Liquid
26053
AUTS2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBRSL2/KIAA0442/MGC13140
AUTS2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LKPQERVEKRQTPLTKKKREALTNGLSFHSKKSRLSHPHHYSSDRENDRNLCQHLGKRKKMPKALRQLKPGQNSCRDSDSESASGESKGFH
RUO
AUTS2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers