Learn More
Abnova™ Human ATP12A Partial ORF (NP_001667, 174 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000479-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the family of P-type cation transport ATPases. This gene encodes a catalytic subunit of the ouabain-sensitive H+/K+ -ATPase that catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. It is also responsible for potassium absorption in various tissues. [provided by RefSeq]
Sequence: SSFNKMIPQQALVIRDSEKKTIPSEQLVVGDIVEVKGGDQIPADIRVLSSQGCRVDNSSLTGESEPQPRSSEFTHENPLETKNICFYSTTCLEASTSPVGTVTGMVINSpecifications
NP_001667 | |
Liquid | |
479 | |
ATP12A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP1AL1 | |
ATP12A | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SSFNKMIPQQALVIRDSEKKTIPSEQLVVGDIVEVKGGDQIPADIRVLSSQGCRVDNSSLTGESEPQPRSSEFTHENPLETKNICFYSTTCLEASTSPVGTVTGMVIN | |
RUO | |
ATP12A | |
Wheat Germ (in vitro) | |
GST |