missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATG10 Full-length ORF (NP_113670.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083734-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105463

  • 508.00€ / 25µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes. ATG10 is an E2-like enzyme involved in 2 ubiquitin-like modifications essential for autophagosome formation: ATG12 (MIM 609608)-ATG5 (MIM 604261) conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A; MIM 601242), a homolog of yeast Apg8, to a membrane-bound form (Nemoto et al., 2003 [PubMed 12890687]).[supplied by OMIM]

Sequence: MEEDEFIGEKTFQRYCAEFIKHSQQIGDSWEWRPSKDCSDGYMCKIHFQIKNGSVMSHLGASTHGQTCLPMEEAFELPLDDCEVIETAAASEVIKYEYHVLYSCSYQVPVLYFRASFLDGRPLTLKDIWEGVHECYKMRLLQGPWDTITQQEHPILGQPFFVLHPCKTNEFMTPVLKNSQKINKNVNYITSWLSIVGPVVGLNLPLSYAKATSQDERNVP
Specifications

Specifications

NP_113670.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ATG10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
83734
ATG10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APG10/APG10L/DKFZp586I0418/FLJ13954/pp12616
ATG10
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers