missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASTN2 Full-length ORF (AAH29272.1, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023245-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157092

  • 359.00€ / 10µg

Estimated Shipment: 29-07-2022
to see stock.
Add to basket

Description

Description

Sequence: MNTLLCKGMFCLLSWEADSRGRLGEYTLQPLSLQTEETTELGSKKELKSMPFITYLSGLLTAQMLSDDQLISGVEIRCEEKGRCPSTCHLCRRPGKEQLSPTPVLLEINRVVPLYTLIQDNGTKEAFKSALMSSYWCSGKGDVIDDWCRCDLSAFDANGLPNCSPLLQPVLRLSPTVEPSSTVVSLEWVDVQPAIGTKVSDYILQHKKVDEYTDTDLYTGEFLSFADDLLSGLGTSCVAAGRSHGEVPEVSIYSVIFKCLEPDGLYKFTLYAVDTRGRHSELSTVTLRTACPLVDDNKAEEIADKIYNLYNGYTSGKEQQMAYNTLMEVSASMLFRVQHHYNSHYEKFGDFVWRSEDELGPRKAHLILRRLERVSSHCSSLLRSAYIQSRVETVPYLFCRSEEVRPAGMVWYSILKDTKIMCEEKMVSMARNTYGESKGR
Specifications

Specifications

AAH29272.1
Liquid
23245
ASTN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0634/bA67K19.1
ASTN2
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ASTN2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers