missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASPH Partial ORF (NP_115857, 161 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000444-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104974

  • 528.00€ / 25µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

This gene is thought to play an important role in calcium homeostasis. The gene is expressed from two promoters and undergoes extensive alternative splicing. The encoded set of proteins share varying amounts of overlap near their N-termini but have substantial variations in their C-terminal domains resulting in distinct functional properties. The longest isoforms (a and f) include a C-terminal Aspartyl/Asparaginyl beta-hydroxylase domain that hydroxylates aspartic acid or asparagine residues in the epidermal growth factor (EGF)-like domains of some proteins, including protein C, coagulation factors VII, IX, and X, and the complement factors C1R and C1S. Other isoforms differ primarily in the C-terminal sequence and lack the hydroxylase domain, and some have been localized to the endoplasmic and sarcoplasmic reticulum. Some of these isoforms are found in complexes with calsequestrin, triadin, and the ryanodine receptor, and have been shown to regulate calcium release from the sarcoplasmic reticulum. Some isoforms have been implicated in metastasis. [provided by RefSeq]

Sequence: PTGEPQQEDDEFLMATDVDDRFETLEPEVSHEETEHSYHVEETVSQDCNQDMEEMMSEQENPDSSEPVVEDERLHHDTDDVTYQVYEEQAVYEPLENEGIEITEVTAPPE
Specifications

Specifications

NP_115857
Liquid
444
ASPH (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAH/CASQ2BP1/HAAH/JCTN/junctin
ASPH
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PTGEPQQEDDEFLMATDVDDRFETLEPEVSHEETEHSYHVEETVSQDCNQDMEEMMSEQENPDSSEPVVEDERLHHDTDDVTYQVYEEQAVYEPLENEGIEITEVTAPPE
RUO
ASPH
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers