missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASH2L Partial ORF (NP_004665.1, 424 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009070-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119865

  • 545.00€ / 25µg

Estimated Shipment: 06-09-2022
to see stock.
Add to basket

Description

Description

Sequence: EITVDEMPPDTAARLGWSQPLGNLQAPLGYDKFSYSWRSKKGTKFHQSIGKHYSSGYGQGDVLGFYINLPEDTETAKSLPDTYKDKALIKFKSYLYFEEKDFVDKAE
Specifications

Specifications

NP_004665.1
Liquid
9070
ASH2L (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASH2/ASH2L1/ASH2L2/Bre2
ASH2L
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EITVDEMPPDTAARLGWSQPLGNLQAPLGYDKFSYSWRSKKGTKFHQSIGKHYSSGYGQGDVLGFYINLPEDTETAKSLPDTYKDKALIKFKSYLYFEEKDFVDKAE
RUO
ASH2L
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers