missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ASCC2 Full-length ORF (NP_115580.2, 1 a.a. - 757 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084164-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195673

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MPALPLDQLQITHKDPKTGKLRTSPALHPEQKADRYFVLYKPPPKDNIPALVEEYLERATFVANDLDWLLALPHDKFWCQVIFDETLQKCLDSYLRYVPRKFDEGVASAPEVVDMQKRLHRSVFLTFLRMSTHKESKDHFISPSAFGEILYNNFLFDIPKILDLCVLFGKGNSPLLQKMIGNIFTQQPSYYSDLDETLPTILQVFSNILQHCGLQGDGANTTPQKLEERGRLTPSDMPLLELKDIVLYLCDTCTTLWAFLDIFPLACQTFQKHDFCYRLASFYEAAIPEMESAIKKRRLEDSKLLGDLWQRLSHSRKKLMEIFHIILNQICLLPILESSCDNIQGFIEEFLQIFSSLLQEKRFLRDYDALFPVAEDISLLQQASSVLDETRTAYILQAVESAWEGVDRRKATDAKDPSVIEEPNGEPNGVTVTAEAVSQASSHPENSEEEECMGAAAAVGPAMCGVELDSLISQVKDLLPDLGEGFILACLEYYHYDPEQVINNILEERLAPTLSQLDRNLDREMKPDPTPLLTSRHNVFQNDEFDVFSRDSVDLSRVHKGKSTRKEENTRSLLNDKRAVAAQRQRYEQYSVVVEEVPLQPGESLPYHSVYYEDEYDDTYDGNQVGANDADSDDELISRRPFTIPQVLRTKVPREGQEEDDDDEEDDADEEAPKPDHFVQDPAVLREKAEARRMAFLAKKGYRHDSSTAVAGSPRGHGQSRETTQERRKKEANKATRANHNRRTMADRKRSKGMIPS
Specifications

Specifications

NP_115580.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ASCC2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
84164
ASCC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASC1p100
ASCC2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers