missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARX Partial ORF (NP_620689, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00170302-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102317

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene is a homeobox-containing gene expressed during development. The expressed protein contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This gene is thought to be involved in CNS development. Mutations in this gene cause X-linked mental retardation and epilepsy. [provided by RefSeq]

Sequence: MSNQYQEEGCSERPECKSKSPTLLSSYCIDSILGRRSPCKMRLLGAAQSLPAPLTSRADPEKAVQGSPKSSSAPFEAELHLPPKLRRLYGPGGGR
Specifications

Specifications

NP_620689
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSNQYQEEGCSERPECKSKSPTLLSSYCIDSILGRRSPCKMRLLGAAQSLPAPLTSRADPEKAVQGSPKSSSAPFEAELHLPPKLRRLYGPGGGR
RUO
ARX
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
170302
ARX (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ISSX/MRX29/MRX32/MRX33/MRX36/MRX38/MRX43/MRX54/MRX76/MRX87/MRXS1/PRTS
ARX
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers