missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARSH Full-length ORF (AAI53086.1, 1 a.a. - 562 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00347527-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190284

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Sulfatases, such as ARSH, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 [PubMed 16174644]).[supplied by OMIM]

Sequence: MTRNARPNIVLLMADDLGVGDLCCYGNNSVSTPNIDRLASEGVRLTQHLAAASMCTPSRAAFLTGRYPIRSGMVSAYNLNRAFTWLGGSGGLPTNETTFAKLLQHRGYRTGLIGKWHLGLSCASRNDHCYHPLNHGFHYFYGVPFGLLSDCQASKTPELHRWLRIKLWISTVALALVPFLLLIPKFARWFSVPWKVIFVFALLAFLFFTSWYSSYGFTRRWNCILMRNHEIIQQPMKEEKVASLMLKEALAFIERYKREPFLLFFSFLHVHTPLISKKKFVGRSKYGRYGDNVEEMDWMVGKILDALDQERLANHTLVYFTSDNGGHLEPLDGAVQLGGWNGIYKGGKGMGGWEGGIRVPGIFRWPSVLEAGRVINEPTSLMDIYPTLSYIGGGILSQDRVIDGQNLMPLLEGRASHSDHEFLFHYCGVYLHTVRWHQKDCATVWKAHYVTPKFYPEGTGACYGSGICSCSGDVTYHDPPLLFDISRDPSEALPLNPDNEPLFDSVIKKMEAAIREHRRTLTPVPQQFSVFNTIWKPWLQPCCGTFPFCGCDKEDDILPMAP
Specifications

Specifications

AAI53086.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.77kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ARSH
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
347527
ARSH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
sulfatase
ARSH
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers