missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARMC4 Partial ORF (NP_060546, 945 a.a. - 1044 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055130-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: AISRCCMWGRNRVAFGEHKAVAPLVRYLKSNDTNVHRATAQALYQLSEDADNCITMHENGAVKLLLDMVGSPDQDLQEAAAGCISNIRRLALATEKARYTSpecifications
NP_060546 | |
Liquid | |
55130 | |
ARMC4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434P1735/FLJ10376/FLJ10817/FLJ32798/RP11-691I13.1 | |
ARMC4 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AISRCCMWGRNRVAFGEHKAVAPLVRYLKSNDTNVHRATAQALYQLSEDADNCITMHENGAVKLLLDMVGSPDQDLQEAAAGCISNIRRLALATEKARYT | |
RUO | |
ARMC4 | |
Wheat Germ (in vitro) | |
GST |