missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARL9 Full-length ORF (NP_996802.1, 1 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00132946-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ARL9 is a member of the small GTPase protein family with a high degree of similarity to ARF (MIM 103180) proteins of the RAS superfamily.[supplied by OMIM]
Sequence: MEFLEIGGSKPFRSYWEMYLSKGLLLIFVVDSADHSRLPEAKKYLHQLIAANPVLPLVVFANKQDLEAAYHITDIHEALALSEVGNDRKMFLFGTYLTKNGSEIPSTMQDAKDLIAQLAADVQSpecifications
NP_996802.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARL9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
132946 | |
ARL9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARL9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |