missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARL13B Partial ORF (NP_878899.1, 329 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00200894-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KNEDETDRPSLESANGKKKTKKLRMKRNHRVEPLNIDDCAPESPTPPPPPPPVGWGTPKVTRLPKLEPLGETHHNDFYRKPLPPLAVPQRPNSDAHDVISSpecifications
NP_878899.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KNEDETDRPSLESANGKKKTKKLRMKRNHRVEPLNIDDCAPESPTPPPPPPPVGWGTPKVTRLPKLEPLGETHHNDFYRKPLPPLAVPQRPNSDAHDVIS | |
RUO | |
ARL13B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
200894 | |
ARL13B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARL2L1/DKFZp686E2075/DKFZp686L2472/DKFZp686M2074/DKFZp761H079/JBTS8/MGC120611/MGC120612 | |
ARL13B | |
Recombinant | |
wheat germ expression system |