Learn More
Abnova™ Human ARHGAP6 Partial ORF (NP_038286, 531 a.a. - 636 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000395-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of actin polymerization at the plasma membrane during several cellular processes. This protein is thought to have two independent functions, one as a GTPase-activating protein with specificity for RhoA, and another as a cytoskeletal protein that promotes actin remodeling. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: RHADDNISKDGQEVTGNKMTSLNLATIFGPNLLHKQKSSDKEFSVQSSARAEESTAIIAVVQKMIENYEALFMVPPDLQNEVLISLLETDPDVVDYLLRRKASQSSSpecifications
NP_038286 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RHADDNISKDGQEVTGNKMTSLNLATIFGPNLLHKQKSSDKEFSVQSSARAEESTAIIAVVQKMIENYEALFMVPPDLQNEVLISLLETDPDVVDYLLRRKASQSS | |
RUO | |
ARHGAP6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
395 | |
ARHGAP6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RHOGAP6/RHOGAPX-1 | |
ARHGAP6 | |
Recombinant | |
wheat germ expression system |