missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARHGAP15 Full-length ORF (AAH38976.2, 1 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055843-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121873

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

RHO GTPases (see ARHA; MIM 165390) regulate diverse biologic processes, and their activity is regulated by RHO GTPase-activating proteins (GAPs), such as ARHGAP15 (Seoh et al., 2003 [PubMed 12650940]).[supplied by OMIM]

Sequence: MQKSTNSDTSVETLNSTRQGTGAVQMRIKNANSHHDRLSQSKSMILTDVGKVTEPISRHRRNHSQHILKDVIPPLEQLMVEKEGYLQKAKIADGGKKLRKNWSTSWIVLSSRRIEFYKESKQQALSNMKTGHKPESVDLCGAHIEWAKEKSSRKNVFQITTVSGNEFLLQSDIDFIILDWFHAIKNAIDRLPKDSSCPSRNLELFKIQRSSSTELLSHYDSDIKEQKPEHRKSLMFRLHHSASDTSDKNRVKSRLKKFITRRPSLKTLQEKGLIKDQIFGSHLHKVCERENSTVPWFVKQCIEAVEKRGLDVDGIYRVSGNLATIQKLRFIVNQEEKLNLDDSQWEDIHVVTGALKMFFRELPEPLFPYSFFEQFVEAIKKQDNNTRIEAVKSLVQKLPPPNRDTMKVLFGHLTKIVAKASKNLMSTQSLGIVFGPTLLRAENETGNMAIHMVYQNQIAELMLSEYSKIFGSEED
Specifications

Specifications

AAH38976.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ARHGAP15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
55843
ARHGAP15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BM046
ARHGAP15
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers