missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARF4L Partial ORF (AAH00043, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000379-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121801

  • 359.00€ / 10µg

Estimated Shipment: 06-09-2022
to see stock.
Add to basket

Description

Description

ADP-ribosylation factor 4D is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4D is closely similar to ARL4A and ARL4C and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in membrane-associated intracellular trafficking. Mutations in this gene have been associated with Bardet-Biedl syndrome (BBS). [provided by RefSeq]

Sequence: MGNHLTEMAPTASSFLPHFQALHVVVIGLDSAGKTSLLYRLKFKEFVQSVPTKGFNTEKIRVPLGGSRGITFQVWDVGGQEKLRPLWRSYTRRTDGLVFV
Specifications

Specifications

AAH00043
Liquid
379
ARF4L (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARF4L/ARL6
ARL4D
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNHLTEMAPTASSFLPHFQALHVVVIGLDSAGKTSLLYRLKFKEFVQSVPTKGFNTEKIRVPLGGSRGITFQVWDVGGQEKLRPLWRSYTRRTDGLVFV
RUO
ARL4D
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers