missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The Human APP ELISA Kit (Colorimetric) measures human APP in cell lysate, cerebral spinal fluid, plasma and serum samples.
- Assay Type: Competitive ELISA
- Standard Curve Range: 11.72 - 1500 pM
- Sensitivity: 0.92 pM
- Inter-Assay: Mean 5.9%
- Intra-Assay: Mean 2.5%
- Spike Recovery:
- Cell Lysate - 101.1%
- Cerebrospinal Fluid - 93.6%
- Sample Volume: 50 μL
- Components:
- Assay Buffer
- Antibody-HRP-conjugate
- Detector Antibody
- Standard
- Microtiter Plate
- TMB Substrate
- Wash Buffer Concentrate
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 351 |
| Gene Symbol | APP |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?