Learn More
R&D Systems™ Human Apolipoprotein E R2 (NP_004622) VersaClone cDNA
Product Code. 18720399
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18720399 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18720399 Supplier R&D Systems™ Supplier No. RDC2616
  • ONLINE EXCLUSIVE PRICE
    773.00€
    730.80€ / 10µg
    Save 42.20€
    5% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Human Apolipoprotein E R2 (NP_004622) VersaClone cDNA clone contains the complete ORF for Apolipoprotein E R2/ApoE R2, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The Apolipoprotein E R2/ApoE R2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifications
Gene hApolipoprotein E R2
Accession Number NP_004622
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 7804
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Quantity 10 μg