missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOL4 Full-length ORF (ENSP00000331089, 1 a.a. - 107 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080832-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125053

  • 508.00€ / 25µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein. [provided by RefSeq]

Sequence: MGSWVQLITSVGTSGLFLGVRVREEGAGMRCSKTIQAGQWLDSSKGPLGPSPPPVPTAGYSSSFCVHYVNLLPGVLVLSVTSQYPHLSMALCQLAAHDWPRLSCVCV
Specifications

Specifications

ENSP00000331089
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
APOL4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
80832
APOL4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOL-IV/APOLIV
APOL4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers