missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOB48R Partial ORF (NP_061160, 71 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055911-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118526

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: RGSQNEGAGRLRGPGDDRRHEVGSSAVEQTWGWGDGSSHGSQAERQDSGAGETAKAARCQEPSAHLEARKKSKAGSGACQDRSGQAQERQESHEQEVNRE
Specifications

Specifications

NP_061160
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RGSQNEGAGRLRGPGDDRRHEVGSSAVEQTWGWGDGSSHGSQAERQDSGAGETAKAARCQEPSAHLEARKKSKAGSGACQDRSGQAQERQESHEQEVNRE
RUO
APOB48R
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
55911
APOB48R (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APOB48R
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers