missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APBB2 Partial ORF (AAH27946, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH27946 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 323 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131661
|
Abnova™
H00000323-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment:
Log in to see stock. |
|||||
16121661
|
Abnova™
H00000323-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment:
Log in to see stock. |
|||||
Description
The protein encoded by this gene interacts with the cytoplasmic domains of amyloid beta (A4) precursor protein and amyloid beta (A4) precursor-like protein 2. This protein contains two phosphotyrosine binding (PTB) domains, which are thought to function in signal transduction. [provided by RefSeq]
Sequence: MSEVLPADSGVDTLAVFMASSGTTDVTNRNSPATPPNTLNLRSSHNELLNAEIKHTETKNSTPPKCRKKYALTNIQAAMGLSDPAAQPLLGNGSANIKLVSpecifications
AAH27946 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434E033/FE65L/FE65L1/MGC35575 | |
APBB2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
323 | |
APBB2 (Human) Recombinant Protein (Q01) | |
MSEVLPADSGVDTLAVFMASSGTTDVTNRNSPATPPNTLNLRSSHNELLNAEIKHTETKNSTPPKCRKKYALTNIQAAMGLSDPAAQPLLGNGSANIKLV | |
RUO | |
APBB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |