missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AP2B1 Full-length ORF (AAH06201, 1 a.a. - 951 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000163-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191321

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is one of two large chain components of the assembly protein complex 2, which serves to link clathrin to receptors in coated vesicles. The encoded protein is found on the cytoplasmic face of coated vesicles in the plasma membrane. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MTDSKYFTTNKKGEIFELKAEPNNEKKEKRKEAVKKVIAAMTVGKDVSSLFPDVVNCMQTDNLELKKLVYLYLMNYAKSQPDMAIMAVNSFVKDCEDPNPLIRALAVRTMGCIRVDKITEYLCEPLRKCLKDEDPYVRKTAAVCVAKLHDINAQMVEDQGFLDSLRDLIADSNPMVVANAVAALSEISESHPNSNLLDLNPQNINKLLTALNECTEWGQIFILDCLSNYNPKDDREAQSICERVTPRLSHANSAVVLSAVKVLMKFLELLPKDSDYYNMLLKKLAPPLVTLLSGEPEVQYVALRNINLIVQKRPEILKQEIKVFFVKYNDPIYVKLEKLDIMIRLASQANIAQVLAELKEYATEVDVDFVRKAVRAIGRCAIKVEQSAERCVSTLLDLIQTKVNYVVQEAIVVIRDIFRKYPNKYESIIATLCENLDSLDEPDARAAMIWIVGEYAERIDNADELLESFLEGFHDESTQVQLTLLTAIVKLFLKKPSETQELVQQVLSLATQDSDNPDLRDRGYIYWRLLSTDPVTAKEVVLSEKPLISEETDLIEPTLLDELICHIGSLASVYHKPPNAFVEGSHGIHRKHLPIHHGSTDAGDSPVGTTTATNLEQPQVIPSQGDLLGDLLNLDLGPPVNVPQVSSMQMGAVDLLGGGLDSLLGSDLGGGIGGSPAVGQSFIPSSVPATFAPSPTPAVVSSGLNDLFELSTGIGMAPGGYVAPKAVWLPAVKAKGLEISGTFTHRQGHIYMEMNFTNKALQHMTDFAIQFNKNSFGVIPSTPLAIHTPLMPNQSIDVSLPLNTLGPVMKMEPLNNLQVAVKNNIDVFYFSCLIPLNVLFVEDGKMERQVFLATWKDIPNENELQFQIKECHLNADTVSSKLQNNNVYTIAKRNVEGQDMLYQSLKLTNGIWILAELRIQPGNPNYTLSLKCRAPEVSQYIYQVYDSILKN
Specifications

Specifications

AAH06201
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
130.35kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AP2B1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
163
AP2B1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADTB2/AP105B/AP2-BETA/CLAPB1/DKFZp781K0743
AP2B1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers