Learn More
Abnova™ Human AOAH Partial ORF (NP_001628, 478 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000313-Q01L
Additional Details : Weight : 0.00010kg
Description
Acyloxyacyl hydrolase (AOAH) is a 2-subunit lipase which selectively hydrolyzes the secondary (acyloxyacyl-linked) fatty acyl chains from the lipid A region of bacterial endotoxins. AOAH may modulate host inflammatory responses to gram-negative bacterial invasion. The 2 subunits are encoded by a single mRNA. [provided by RefSeq]
Sequence: EQLSNTLKKIAASEKFTNFNLFYMDFAFHEIIQEWQKRGGQPWQLIEPVDGFHPNEVALLLLADHFWKKVQLQWPQILGKENPFNPQIKQVFGDQGGHSpecifications
NP_001628 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EQLSNTLKKIAASEKFTNFNLFYMDFAFHEIIQEWQKRGGQPWQLIEPVDGFHPNEVALLLLADHFWKKVQLQWPQILGKENPFNPQIKQVFGDQGGH | |
RUO | |
AOAH | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
313 | |
AOAH (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AOAH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |