Learn More
Abnova™ Human ANTXR1 Full-length ORF (NP_444262.1, 1 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084168-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colorectal cancer. The encoded protein has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax. The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to this receptor protein mediates delivery of toxin components to the cytosol of cells. Once inside the cell, the other two components of anthrax toxin, edema factor (EF) and lethal factor (LF) disrupt normal cellular processes. Three alternatively spliced variants that encode different protein isoforms have been described. [provided by RefSeq]
Sequence: MATAERRALGIGFQWLSLATLVLICAGQGGRREDGGPACYGGFDLYFILDKSGSVLHHWNEIYYFVEQLAHKFISPQLRMSFIVFSTRGTTLMKLTEDREQIRQGLEELQKVLPGGDTYMHEGFERASEQIYYENRQGYRTASVIIALTDGELHEDLFFYSEREANRSRDLGAIVYCVGVKDFNETQLARIADSKDHVFPVNDGFQALQGIIHSILKKSCIEILAAEPSTICAGESFQVVVRGNGFRHARNVDRVLCSFKINDSVTLNEKPFSVEDTYLLCPAPILKEVGMKAALQVSMNDGLSFISSSVIITTTHCSDGSILAIALLILFLLLALALLWWFWPLCCTVIIKEVPPPPAEESEENKIKSpecifications
NP_444262.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ANTXR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84168 | |
ANTXR1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATR/FLJ10601/FLJ11298/FLJ21776/TEM8 | |
ANTXR1 | |
Recombinant | |
wheat germ expression system |