missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANKRD27 Full-length ORF (NP_115515.2, 1 a.a. - 1050 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084079-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105623

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MALYDEDLLKNPFYLALQKCRPDLCSKVAQIHGIVLVPCKGSLSSSIQSTCQFESYILIPVEEHFQTLNGKDVFIQGNRIKLGAGFACLLSVPILFEETFYNEKEESFSILCIAHPLEKRESSEEPLAPSDPFSLKTIEDVREFLGRHSERFDRNIASFHRTFRECERKSLRHHIDSANALYTKCLQQLLRDSHLKMLAKQEAQMNLMKQAVEIYVHHEIYNLIFKYVGTMEASEDAAFNKITRSLQDLQQKDIGVKPEFSFNIPRAKRELAQLNKCTSPQQKLVCLRKVVQLITQSPSQRVNLETMCADDLLSVLLYLLVKTEIPNWMANLSYIKNFRFSSLAKDELGYCLTSFEAAIEYIRQGSLSAKPPESEGFGDRLFLKQRMSLLSQMTSSPTDCLFKHIASGNQKEVERLLSQEDHDKDTVQKMCHPLCFCDDCEKLVSGRLNDPSVVTPFSRDDRGHTPLHVAAVCGQASLIDLLVSKGAMVNATDYHGATPLHLACQKGYQSVTLLLLHYKASAEVQDNNGNTPLHLACTYGHEDCVKALVYYDVESCRLDIGNEKGDTPLHIAARWGYQGVIETLLQNGASTEIQNRLKETPLKCALNSKILSVMEAYHLSFERRQKSSEAPVQSPQRSVDSISQESSTSSFSSMSASSRQEETKKDYREVEKLLRAVADGDLEMVRYLLEWTEEDLEDAEDTVSAADPEFCHPLCQCPKCAPAQKRLAKVPASGLGVNVTSQDGSSPLHVAALHGRADLIPLLLKHGANAGARNADQAVPLHLACQQGHFQVVKCLLDSNAKPNKKDLSGNTPLIYACSGGHHELVALLLQHGASINASNNKGNTALHEAVIEKHVFVVELLLLHGASVQVLNKRQRTAVDCAEQNSKIMELLQVVPSCVASLDDVAETDRKEYVTVKIRKKWNSKLYDLPDEPFTRQFYFVHSAGQFKGKTSREIMARDRSVPNLTEGSLHEPGRQSVTLRQNNLPAQSGSHAAEKGNSDWPERPGLTQTGPGHRRMLRRHTVEDAVVSQGPEAAGPLSTPQEVSASRS
Specifications

Specifications

NP_115515.2
Liquid
84079
ANKRD27 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434L0718/FLJ00040/PP12899/VARP
ANKRD27
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
143.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ANKRD27
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers