missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANKRD21 Partial ORF (NP_778146.1, 485 a.a. - 584 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00317754-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192777

  • 349.00€ / 10µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: NTGISQDEILTNKQKQIEVAEQKMNSELSLSHKKEEDLLRENSVLQEEIAVLRLELDETKHQNQLRENKILEEIESVKEKTDKLLRAMQLNEEALTKTNI
Specifications

Specifications

NP_778146.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NTGISQDEILTNKQKQIEVAEQKMNSELSLSHKKEEDLLRENSVLQEEIAVLRLELDETKHQNQLRENKILEEIESVKEKTDKLLRAMQLNEEALTKTNI
RUO
POTED
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
317754
ANKRD21 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A26B3/ANKRD21/POTE/POTE-21/POTE21
POTED
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers