missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANKRD15 Partial ORF (AAH38116, 701 a.a. - 800 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023189-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107032

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein containing four ankyrin repeat domains in its C-terminus. The suggested role for this protein is in tumorigenesis of renal cell carcinoma. Two alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MGSLNSQLISTLSSINSVMKSASTEELRNPDFQKTSLGKITGNYLGYTCKCGGLQSGSPLSSQTSQPEQEVGTSEGKPISSLDAFPTQEGTLSPVNLTDD
Specifications

Specifications

AAH38116
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSLNSQLISTLSSINSVMKSASTEELRNPDFQKTSLGKITGNYLGYTCKCGGLQSGSPLSSQTSQPEQEVGTSEGKPISSLDAFPTQEGTLSPVNLTDD
RUO
KANK1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23189
ANKRD15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANKRD15/DKFZp451G231/KANK/KIAA0172/MGC43128
KANK1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers