missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Angiopoietin-like 4 Biotin, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ BT4487-050
This item is not returnable.
View return policy
Description
The Recombinant Human Angiopoietin-like 4 Biotin Protein is derived from NS0. The Recombinant Human Angiopoietin-like 4 Biotin Protein has been validated for the following applications: Bioactivity.Specifications
| Q9BY76 | |
| Theoretical M.W.: 44kDa (unlabeled) | |
| Mouse myeloma cell line,NS0-derived human Angiopoietin-like Protein 4/ANGPTL4 protein Gly26-Ser406 (Lys163Ala,Arg164Ala),with a C-terminal 6-His tag | |
| <1.0 EU per 1μg of the protein by the LAL method. | |
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Measured by its ability to promote the expansion of E16 rat liver mononuclear cells in vitro, in the presence of Recombinant Mouse SCF/c-kit Ligand, Recombinant Mouse Thrombopoietin/Tpo, and Recombinant Mouse Flt-3 Ligand. The ED50 for this effect is typically 100-600ng/mL in the presence of a cross-linking antibody, His Tag MAb, Mouse Anti-polyHistidine Monoclonal Antibody. | |
| Yes |
| 51129 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| ANGPTL4 | |
| Biotin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?