missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANAPC10 Partial ORF (AAH05217, 70 a.a. - 185 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010393-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194992

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: RKTTVKTLCIYADYKSDESYTPSKISVRVGNNFHNLQEIRQLELVEPSGWIHVPLTDNHKKPTRTFMIQIAVLANHQNGRDTHMRQIKIYTPVEESSIGKFPRCTTIDFMMYRSIR
Specifications

Specifications

AAH05217
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.39kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RKTTVKTLCIYADYKSDESYTPSKISVRVGNNFHNLQEIRQLELVEPSGWIHVPLTDNHKKPTRTFMIQIAVLANHQNGRDTHMRQIKIYTPVEESSIGKFPRCTTIDFMMYRSIR
RUO
ANAPC10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10393
ANAPC10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APC10/DKFZp564L0562/DOC1
ANAPC10
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers