missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AMICA1 Full-length ORF (AAI00798.1, 1 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00120425-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127483

  • 335.00€ / 10µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MGCVFQSTEDKCIFKIDWTLSPGEHAKDEYVLYYYSNLSVPIGRFQNRVHLMGDILCNDGSLLLQDVQEADQGTYICEIRLKGESQVFKKAVVLHVLPEEPKELMVHVGGLIQMGCVFQSTEVKHVTKVEWIFSGRRAKEEIVFRYYHKLRMSVEYSQSWGHFQNRVNLVGDIFRNDGSIMLQGVRESDGGNYTCSIHLGNLVFKKTIVLHVSPEEPRTLVTPAALRPLVLGGNQLVIIVGIVCATILLLPVLILIVKKTCGNKSSVNSTVLVKNTKKTNPEIKEKPCHFERCEGEKHIYSPIIVREVIEEEEPSEKSEATYMTMHPVWPSLRSDRNNSLEKKSGGGMPKTQQAF
Specifications

Specifications

AAI00798.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AMICA1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
120425
AMICA1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMICA/CREA7-1/CREA7-4/FLJ37080/Gm638/JAML/MGC118814/MGC118815
AMICA1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers