Learn More
Abnova™ Human ALPI Partial ORF (NP_001622, 74 a.a. - 162 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000248-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation. [provided by RefSeq]
Sequence: LKGQKNGKLGPETPLAMDRFPYLALSKTYNVDRQVPDSAATATAYLCGVKANFQTIGLSAAARFNQCNTTRGNEVISVMNRAKQAGKSVSpecifications
NP_001622 | |
Liquid | |
248 | |
ALPI (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IAP | |
ALPI | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LKGQKNGKLGPETPLAMDRFPYLALSKTYNVDRQVPDSAATATAYLCGVKANFQTIGLSAAARFNQCNTTRGNEVISVMNRAKQAGKSV | |
RUO | |
ALPI | |
Wheat Germ (in vitro) | |
GST |