missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGTRAP Full-length ORF (NP_001035284.1, 1 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057085-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102463

  • 528.00€ / 25µg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a transmembrane protein localized to the plasma membrane and perinuclear vesicular structures. The gene product interacts with the angiotensin II type I receptor and negatively regulates angiotensin II signaling. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MELPAVNLKVILLGHWLLTTWGCIVFSGSYAWANFTILALGVWAVAQRDSIDAISMFLGGLLATIFLDIVHISIFYPRVSLTDTGRFGVGMAILSLLLKPLSCCFVYHMYRERGGFLGSSQDRSAYQTIDSAEAPADPFAVPEGRSQDARGY
Specifications

Specifications

NP_001035284.1
Liquid
57085
AGTRAP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATRAP/MGC29646
AGTRAP
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AGTRAP
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers