missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AGR2 Partial ORF (AAH15503.1, 61 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010551-Q03.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: ALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLVYETTDKHLSPDGQYVPRIMFVDPSLTVRADITGRYSNRLYAYEPADTALLSpecifications
AAH15503.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ALYKSKTSNKPLMIIHHLDECPHSQALKKVFAENKEIQKLAEQFVLLNLVYETTDKHLSPDGQYVPRIMFVDPSLTVRADITGRYSNRLYAYEPADTALL | |
RUO | |
AGR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10551 | |
AGR2 (Human) Recombinant Protein (Q03) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AG2/GOB-4/HAG-2/XAG-2 | |
AGR2 | |
Recombinant | |
wheat germ expression system |