Learn More
Abcam Human AGO2 knockout HeLa cell line
Description
AGO2 KO cell line available to order. KO validated. Free of charge wild type control available. Homozygote. 338 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 1,000,000 Cells |
| Regulatory Status | RUO |