Learn More
Abcam Human AGO2 knockout HeLa cell line
Product Code. 32154218
Change view
Click to view available options
1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32154218 1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32154218 Supplier Abcam Supplier No. ab3084881000000Cells_
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AGO2 KO cell line available to order. KO validated. Free of charge wild type control available. Homozygote. 338 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
AGO2 KO cell line available to order. KO validated. Free of charge wild type control available. Homozygote. 338 bp deletion. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells