Learn More
R&D Systems™ Human AGK (NP_060708.1) VersaClone cDNA
Specifications
Specifications
| Gene | Acylglycerol Kinase |
| Accession Number | NP_060708.1 |
| Gene ID (Entrez) | 55750 |
| Gene Alias | acylglycerol kinase, EC 2.7.1.107, EC 2.7.1.94, FLJ10842, hAGK, HsMuLK, mitochondrial, MuLK, MULKhsMuLK, Multi-substrate lipid kinase |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |