Learn More
Proteintech™ Human AFP ELISA Kit
Product Code. 16891536 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Product Code. Target Species unitSize
16891536 Human 96 tests
1 options
Product Code. 16891536
Brand: Proteintech™ KE0013296T
Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous AFP
  • Recognizes human AFP
  • AFP (alpha-fetoprotein) is a major plasma protein found in fetus while plasma levels decrease rapidly after birth
    • Early marker of hepatocyte lineage
    • High concentrations correlated with tumor cell growth
    • Detection of AFP important to diagnose hepatocellular carcinoma, stomach cancer and germ cell cancers
    • Altered levels of fetal and maternal AFP associated with hypothyroidism, autoimmune disorders and heart defects
Process:
  • Antibody specific for AFP pre-coated onto microwells
  • AFP protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for AFP is added to detect captured AFP protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Accession Number BC027881
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 0.1 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 174
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P02771
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human AFP ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine