Learn More
Abnova™ Human ADRBK2 Partial ORF (AAH36797, 48 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000157-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the beta adrenergic receptor kinase 2 has 85% amino acid similarity with beta adrenergic receptor kinase 1, with the protein kinase catalytic domain having 95% similarity. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function. [provided by RefSeq]
Sequence: AERNEITFDKIFNQKIGFLLFKDFCLNEINEAVPQVKFYEEIKEYEKLDNEEDRLCRSRQIYDAYIMKELLSCSHPFSKQAVEHVQSHLSKKQVTSTLFQSpecifications
AAH36797 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AERNEITFDKIFNQKIGFLLFKDFCLNEINEAVPQVKFYEEIKEYEKLDNEEDRLCRSRQIYDAYIMKELLSCSHPFSKQAVEHVQSHLSKKQVTSTLFQ | |
RUO | |
ADRBK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
157 | |
ADRBK2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BARK2/GRK3 | |
ADRBK2 | |
Recombinant | |
wheat germ expression system |