missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADRA1A Partial ORF (NP_150646, 1 a.a. - 27 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000148-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194424

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1A-adrenergic receptor. Alternative splicing of this gene generates four transcript variants, which encode four different isoforms with distinct C-termini but having similar ligand binding properties. [provided by RefSeq]

Sequence: MVFLSGNASDSSNCTQPPAPVNISKAI
Specifications

Specifications

NP_150646
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
28.71kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVFLSGNASDSSNCTQPPAPVNISKAI
RUO
ADRA1A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
148
ADRA1A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADRA1C/ADRA1L1/ALPHA1AAR
ADRA1A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers