missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADCY7 Partial ORF (NP_001105, 197 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000113-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194164

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterized by the presence of twelve membrane-spanning domains in its sequences. [provided by RefSeq]

Sequence: HKHQMQDASRDLFTYTVKCIQIRRKLRIEKRQQENLLLSVLPAHISMGMKLAIIERLKEHGDRRCMPDNNFHSLYVKRHQN
Specifications

Specifications

NP_001105
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HKHQMQDASRDLFTYTVKCIQIRRKLRIEKRQQENLLLSVLPAHISMGMKLAIIERLKEHGDRRCMPDNNFHSLYVKRHQN
RUO
ADCY7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
113
ADCY7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AC7/FLJ36387/KIAA0037
ADCY7
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers