missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAM33 Full-length ORF (AAH62663.1, 1 a.a. - 85 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080332-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104973

  • 349.00€ / 10µg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MGWRPRRARGTPLLLLLLLLLLWPVPGAGVLQGHIPGQPVTPHWVLDGQPWRTVSLEEPVSKPDMGLVALEAEGQELLLELEKNQ
Specifications

Specifications

AAH62663.1
Liquid
80332
ADAM33 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C20orf153/DJ964F7.1/DKFZp434K0521/FLJ35308/FLJ36751/MGC149823/MGC71889
ADAM33
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ADAM33
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers