missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACTR6 Full-length ORF (NP_071941.1, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064431-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103633

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MTTLVLDNGAYNAKIGYSHENVSVIPNCQFRSKTARLKTFTANQIDEIKDPSGLFYILPFQKGYLVNWDVQRQVWDYLFGKEMYQVDFLDTNIIITEPYFNFTSIQESMNEILFEEYQFQAVLRVNAGALSAHRYFRDNPSELCCIIVDSGYSFTHIVPYCRSKKKKEAIIRINVGGKLLTNHLKEIISYRQLHVMDETHVINQVKEDVCYVSQDFYRDMDIAKLKGEENTVMIDYVLPDFSTIKKGFCKPREEMVLSGKYKSGEQILRLANERFAVPEILFNPSDIGIQEMGIPEAIVYSIQNLPEEMQPHFFKNIVLTGGNSLFPGFRDRVYSEVRCLTPTDYDVSVVLPENPITYAWEGGKLISENDDFEDMVVTREDYEENGHSVCEEKFDI
Specifications

Specifications

NP_071941.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ACTR6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
64431
ACTR6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARP6/CDA12/FLJ13433/HSPC281/MSTP136/hARP6/hARPX
ACTR6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers