Learn More
Abnova™ Human ACSS2 Full-length ORF (AAH98422.1, 1 a.a. - 606 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055902-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a cytosolic enzyme that catalyzes the activation of acetate for use in lipid synthesis and energy generation. The protein acts as a monomer and produces acetyl-CoA from acetate in a reaction that requires ATP. Expression of this gene is regulated by sterol regulatory element-binding proteins, transcription factors that activate genes required for the synthesis of cholesterol and unsaturated fatty acids. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MKGATTNICYNVLDRNVHEKKLGDKVAFYWEGNEPGETTQITYHQLLVQVCQFSNVLRKQGIQKGDRVAIYMPMIPELVVAMLACARIGALHSIVFAGFSSESLCERILDSSCSLLITTDAFYRGEKLVNLKELADEALQKCQEKGFPVRCCIVVKHLGRAELGMGDSTSQSPPIKRSCPDVQISWNQGIDLWWHELMQEAGDECEPEWCDAEDPLFILYTSGSTGKPKGVVHTVGGYMLYVATTFKYVFDFHAEDVFWCTADIGWITGHSYVTYGPLANGATSVLFEGIPTYPDVNRLWSIVDKYKVTKFYTAPTAIRLLMKFGDEPVTKHSRASLQVLGTVGEPINPEAWLWYHRVVGAQRCPIVDTFWQTETGGHMLTPLPGATPMKPGSATFPFFGVAPAILNESGEELEGEAEGYLVFKQPWPGIMRTVYGNHERFETTYFKKFPGYYVTGDGCQRDQDGYYWITGRIDDMLNVSGHLLSTAEVESALVEHEAVAEAAVVGHPHPVKGECLYCFVTLCDGHTFSPKLTEELKKQIREKIGPIATPDYIQNAPGLPKTRSGKIMRRVLRKIAQNDHDLGDMSTVADPSVISHLFSHRCLTIQSpecifications
AAH98422.1 | |
Liquid | |
55902 | |
ACSS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAS2/ACS/ACSA/AceCS/DKFZp762G026/dJ1161H23.1 | |
ACSS2 | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ACSS2 | |
Wheat Germ (in vitro) | |
GST |