Learn More
Abnova™ Human ABTB1 Full-length ORF (NP_115937.1, 1 a.a. - 336 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080325-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with an ankyrin repeat region and two BTB/POZ domains, which are thought to be involved in protein-protein interactions. Expression of this gene is activated by the phosphatase and tensin homolog, a tumor suppressor. Alternate splicing results in three transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MLDTKWKGKSVVVLRHPLINPVAFGALLQYLYTGRLDIGVEHVSDCERLAKQCQLWDLLSDLEAKCEKVSEFVASKPGTCVKVLTIEPPPADPRLREDMALLADCALPPELRGDLWELPFPCPDGFNSCPDICFRVAGCSFLCHKAFFCGRSDYFRALLDDHFRESEEPATSGGPPAVTLHGISPDVFTHVLYYMYSDHTELSPEAAYDVLSVADMYLLPGLKRLCGRSLAQMLDEDTVVGVWRVAKLFRLARLEDQCTEYMAKVIEKLVEREDFVEAVKEEAAAVAARQETDSIPLVDDIRFHVASTVQTYSAIEEAQQRLRALEDLLVSIGLDCSpecifications
NP_115937.1 | |
Liquid | |
80325 | |
ABTB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BPOZ/Btb3/EF1ABP/MGC20585/PP2259 | |
ABTB1 | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ABTB1 | |
Wheat Germ (in vitro) | |
GST |