Learn More
Abnova™ Human ABCC6 Partial ORF (NP_001162, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000368-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq]
Sequence: IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRVSpecifications
NP_001162 | |
Liquid | |
368 | |
ABCC6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC34/ARA/EST349056/MLP1/MOATE/MRP6/PXE/PXE1 | |
ABCC6 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IAEMGSYQELLQRKGALVCLLDQARQPGDRGEGETEPGTSTKDPRGTSAGRRPELRRERSIKSVPEKDRTTSEAQTEVPLDDPDRAGWPAGKDSIQYGRV | |
RUO | |
ABCC6 | |
Wheat Germ (in vitro) | |
GST |