missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ABCA8 Full-length ORF (AAH47765.1, 1 a.a. - 266 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010351-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104932

  • 528.00€ / 25µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. The function of this protein has not yet been determined. [provided by RefSeq]

Sequence: MRKRKISVCQQTWALLCKNFLKKWRMKRESLMEWLNSLLLLLCLYIYPHSHQVNDFSSLLTMDLGRVDTFNESRFSVVYTPVTNTTQQIMNKVASTPFLAGKEVLGLPDEESIKEFTANYPEEIVRVTFTNTYSYHLKFLLGHGMPAKKEHKDHTAHCYETNEDVYCEVSVFWKEGFVALQAAINAAIIEITTNHSVMEELMSVTGKNMKMHSFIGQSGVITDLYLFSCIISFSSFIYYASVNVTRERKRMKALMTMMGLRDSAFW
Specifications

Specifications

AAH47765.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ABCA8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
10351
ABCA8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0822/MGC163152
ABCA8
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers